2,3-Diaminopyridine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Pyridine-2,3-diyldiamine

M-Clarity™ quality level = MQ200

8.41183.0005 8.41183.0025 8.41183.0005 8.41183.0025
EM8.41183.0005CS 112513.38 CAD
EM8.41183.0005 EM8.41183.0025 CA8.41183.0005 CA8.41183.0025
2,3-Diaminopyridine for synthesis, Sigma-Aldrich®
2,3-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 148…150 °C (5 mmHg)
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006319
CAS Number: 452-58-4
EINECS: 207-200-9
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Melting Point 108 - 113 °C
Molar Mass 109.13 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR