N-Methyl-N-(trimethylsilyl)trifluoroacetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide

M-Clarity™ quality level = MQ200

8.41102.0005 8.41102.0025 8.41102.0005 8.41102.0025
EM8.41102.0005CS 12147.04 CAD
EM8.41102.0005 EM8.41102.0025 CA8.41102.0005 CA8.41102.0025
N-Methyl-N-(trimethylsilyl)trifluoroacetamide for synthesis, Sigma-Aldrich®
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Formula: C₆H₁₂F₃NOSi
MW: 199.25 g/mol
Boiling Pt: 130 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00000411
CAS Number: 24589-78-4
EINECS: 246-331-6
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Molar Mass 199.25 g/mol
Boiling Point 130 - 132 °C (1013 hPa)
Flash Point 25 °C
Density 1.085 g/cm3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR