2-Methyl-3-buten-2-ol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 3-Hydroxy-3-methyl-1-butene

M-Clarity™ quality level = MQ200

8.41089.0250 8.41089.1000 8.41089.0250 8.41089.1000
EM8.41089.0250CS 802.64 CAD
EM8.41089.0250 EM8.41089.1000 CA8.41089.0250 CA8.41089.1000
2-Methyl-3-buten-2-ol for synthesis, Sigma-Aldrich®
2-Methyl-3-buten-2-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: –15 °C
Density: 0.824 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004470
CAS Number: 115-18-4
EINECS: 204-068-4
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -28 °C
Molar Mass 86.13 g/mol
Boiling Point 98 - 99 °C (1013 hPa)
Vapor Pressure 137 hPa (50 °C)
Flash Point 11 °C
Explosion Limit 1.5 - 9.4 %(V)
Density 0.824 g/cm3 (20 °C)
pH 9 - 10 (100 g/l, H2O, 20 °C)
Ignition Point 380 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR