Acetaldehyde oxime (mixture of syn and anti) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.41085.0050 8.41085.0250 8.41085.0050 8.41085.0250
EM8.41085.0050CS 12600.5 CAD
EM8.41085.0050 EM8.41085.0250 CA8.41085.0050 CA8.41085.0250
Acetaldehyde oxime (mixture of syn and anti) for synthesis, Sigma-Aldrich®
Acetaldehyde oxime (mixture of syn and anti)
Formula: C₂H₅NO
MDL Number: MFCD00002124
CAS Number: 107-29-9
EINECS: 203-479-6
UN: 2332
ADR: 3,III

Order Now

Specification Test Results

Solubility 185 g/l (25 °C)
Molar Mass 59.07 g/mol
Boiling Point 115 °C
Vapor Pressure 13 hPa (25 °C)
Flash Point 40 °C
Explosion Limit 4.2 - 50 %(V)
Density 0.969 g/cm3 (20 °C)
Ignition Point 380 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR