Octadecylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.41029.0250 8.41029.1000 8.41029.0250 8.41029.1000
EM8.41029.0250CS 1799.62 CAD
EM8.41029.0250 EM8.41029.1000 CA8.41029.0250 CA8.41029.1000
Octadecylamine for synthesis, Sigma-Aldrich®
Octadecylamine
Formula: H₃C(CH₂)₁₆CH₂NH₂
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 50…52 °C
Density: 0.8618 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008159
CAS Number: 124-30-1
EINECS: 204-695-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 50 - 52 °C
Molar Mass 269.51 g/mol
Boiling Point 349 °C
Vapor Pressure  - 100 hPa (20 °C)
Flash Point 110 °C
Density 0.86 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR