Span® 85 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.40124.0100 8.40124.0500 8.40124.0100 8.40124.0500
EM8.40124.0100CS 1474.14 CAD
EM8.40124.0100 EM8.40124.0500 CA8.40124.0100 CA8.40124.0500
Span® 85 for synthesis, Sigma-Aldrich®
Span® 85
Formula: C₆₀H₁₀₈O₈
Boiling Pt: ∼100 °C (1013 hPa)
Density: 0.95 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00133820
CAS Number: 26266-58-0
EINECS: 247-569-3

Order Now

Specification Test Results

Solubility (25 °C) insoluble
Molar Mass 957.51 g/mol
Boiling Point 100 °C
Vapor Pressure  - 1.4 hPa (20 °C)
Flash Point 149 °C
Density 0.95 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR