Span® 20 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Sorbitan monododecanoate, Arlacel 20, Sorbitan monolaurate

M-Clarity™ quality level = MQ200

8.40119.0100 8.40119.0500 8.40119.0100 8.40119.0500
EM8.40119.0100CS 1129.7 CAD
EM8.40119.0100 EM8.40119.0500 CA8.40119.0100 CA8.40119.0500
Span® 20 for synthesis, Sigma-Aldrich®
Span® 20
Formula: C₁₈H₃₄O₆
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005365
CAS Number: 1338-39-2
EINECS: 215-663-3
Merck Index: 12,08872

Order Now

Specification Test Results

Molar Mass 346.47 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure  - 1.3 hPa (20 °C)
Flash Point 149 °C
Density 1.0 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR