1,5-Naphthalenedisulfonic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.40104.0025 8.40104.0250 8.40104.0025 8.40104.0250
EM8.40104.0025CS 5740.14 CAD
EM8.40104.0025 EM8.40104.0250 CA8.40104.0025 CA8.40104.0250
1,5-Naphthalenedisulfonic acid for synthesis, Sigma-Aldrich®
1,5-Naphthalenedisulphonic acid
Formula: C₁₀H₈O₆S₂
Melting Pt: 110 °C
MDL Number: MFCD00003996
CAS Number: 81-04-9
EINECS: 201-317-9
UN: 2585
ADR: 8,III

Order Now

Specification Test Results

Solubility 1030 g/l (20 °C)
Melting Point 110 °C decomposes
Molar Mass 288.3 g/mol
Bulk Density 900 kg/m3
Vapor Pressure 51 hPa (50 °C)
pH 2.4 (1 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR