2-Dimethylaminoethyl methacrylate stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: DMA, N,N-Dimethylaminoethyl methacrylate

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.40083.0025 8.40083.0250 8.40083.1000 8.40083.0025 8.40083.0250 8.40083.1000
EM8.40083.0025CS 4396.97 CAD
EM8.40083.0025 EM8.40083.0250 EM8.40083.1000 CA8.40083.0025 CA8.40083.0250 CA8.40083.1000
2-Dimethylaminoethyl methacrylate stabilised for synthesis, Sigma-Aldrich®
2-Dimethylaminoethyl methacrylate
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
MDL Number: MFCD00008589
CAS Number: 2867-47-2
EINECS: 220-688-8
UN: 2522
ADR: 6.1 II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -50 °C
Molar Mass 157.21 g/mol
Boiling Point 182 - 192 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 65 °C
Explosion Limit 1.20 %(V)
Density 0.93 g/cm3 (20 °C)
Ignition Point 255 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR