Tetradecyltrimethylammonium bromide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.40006.0025
8.40006.0100
8.40006.0025
8.40006.0100
EM8.40006.0025CS
4493.52
CAD
EM8.40006.0025
EM8.40006.0100
CA8.40006.0025
CA8.40006.0100
Tetradecyltrimethylammonium bromide for synthesis, Sigma-Aldrich®
Tetradecyltrimethylammonium bromide
Formula:
C₁₇H₃₈BrN Melting Pt: 245…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011770 CAS Number: 1119-97-7 EINECS: 214-291-9 UN: 3077 ADR: 9,III Merck Index: 12,06419 |
Specification Test Results
Solubility | 100 g/l (20 °C) |
Melting Point | 245 - 250 °C |
Molar Mass | 336.4 g/mol |
Bulk Density | 600 kg/m3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...