Acetylacetonato(1,5-cyclooctadiene)rhodium(I) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: (Acetylacetonato)(1,5-cyclooctadiene)rhodium(I)

M-Clarity™ quality level = MQ200

8.24420.0100 8.24420.0500 8.24420.0100 8.24420.0500
EM8.24420.0100CS 109369.18 CAD
EM8.24420.0100 EM8.24420.0500 CA8.24420.0100 CA8.24420.0500
Acetylacetonato(1,5-cyclooctadiene)rhodium(I) for synthesis, Sigma-Aldrich®
Acetylacetonato(1,5-cyclooctadiene)rhodium(I)
Formula: C₁₃H₂₀O₂Rh
MDL Number: MFCD00075046
CAS Number: 12245-39-5

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 138 - 140 °C
Molar Mass 310.2 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR