Acetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.22343.0100 8.22343.1000 8.22343.0100 8.22343.1000
EM8.22343.0100CS 1240.3 CAD
EM8.22343.0100 EM8.22343.1000 CA8.22343.0100 CA8.22343.1000
Acetamide for synthesis, Sigma-Aldrich®
Acetamide
Formula: C₂H₅NO
Boiling Pt: 221.1 °C (1013 hPa)
Melting Pt: 78…81 °C
Density: 1.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008023
CAS Number: 60-35-5
EINECS: 200-473-5
Merck Index: 13,00044

Order Now

Specification Test Results

Solubility 2200 g/l (20 °C)
Melting Point 78 - 81 °C
Molar Mass 59.06 g/mol
Boiling Point 221 - 222 °C (1013 hPa)
Vapor Pressure 1.33 hPa (65 °C)
Density 1.159 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR