Paracetamol ≥99.0% (by HPLC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP

M-Clarity™ quality level = MQ200

8.22325.0250 8.22325.1000 8.22325.9025 8.22325.0250 8.22325.1000 8.22325.9025
EM8.22325.0250CS 1993.96 CAD
EM8.22325.0250 EM8.22325.1000 EM8.22325.9025 CA8.22325.0250 CA8.22325.1000 CA8.22325.9025
Paracetamol ≥99.0% (by HPLC) for synthesis, Sigma-Aldrich®
Paracetamol
Formula: C₈H₉NO₂
Boiling Pt: 388 °C (1013 hPa)
Melting Pt: 169…170 °C
Density: 1.293 g/cm3 at 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00002328
CAS Number: 103-90-2
EINECS: 203-157-5
Merck Index: 13,00048

Order Now

Specification Test Results

Solubility 14 g/l (20 °C)
Melting Point 169 - 171 °C
Molar Mass 151.16 g/mol
Boiling Point 500 °C
Explosion Limit 15 %(V)
Density 1.293 g/cm3 (21 °C)
pH 5.5 - 6.5 (H2O, 20 °C) (saturated solution)
Ignition Point 540 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR