Benzotriazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole

M-Clarity™ quality level = MQ200

8.22315.0100 8.22315.0500 8.22315.1000 8.22315.9025 8.22315.0100 8.22315.0500 8.22315.1000 8.22315.9025
EM8.22315.0100CS 1489.94 CAD
EM8.22315.0100 EM8.22315.0500 EM8.22315.1000 EM8.22315.9025 CA8.22315.0100 CA8.22315.0500 CA8.22315.1000 CA8.22315.9025
Benzotriazole for synthesis, Sigma-Aldrich®
Benzotriazole
Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 99 °C
Density: 1.36 g/cm3 at 20 °C
Flash Pt: 170 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005699
CAS Number: 95-14-7
EINECS: 202-394-1
UN: 3077
ADR: 9,III
Merck Index: 13,01109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba54cc42ef5ad9dbae6d7f118a3af1ae642287&vid=ce57b3f0d2583e32220d1e191d0202ad452e6475&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 19 g/l (20 °C)
Melting Point 99 °C
Molar Mass 119.12 g/mol
Bulk Density 500 kg/m3
Boiling Point 350 °C (1013 hPa)
Vapor Pressure 0.053 hPa (20 °C)
Flash Point 212 °C
Density 1.36 g/cm3 (20 °C)
pH 6.0 - 7.0 (100 g/l, H2O, 20 °C) suspension
Ignition Point 400 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR