2-Naphthol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Hydroxynaphthalene, β-Naphthol, β-Hydroxynaphthalene

M-Clarity™ quality level = MQ200

8.22290.0250 8.22290.1000 8.22290.0250 8.22290.1000
EM8.22290.0250CS 489.8 CAD
EM8.22290.0250 EM8.22290.1000 CA8.22290.0250 CA8.22290.1000
2-Naphthol for synthesis, Sigma-Aldrich®
2-Naphthol
Formula: C₁₀H₈O
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 121.6 °C
Density: 1.217 g/cm³ (20 °C)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00004067
CAS Number: 135-19-3
EINECS: 205-182-7
UN: 3077
ADR: 9,III
Merck Index: 12,06471

Order Now

Specification Test Results

Solubility 1 g/l (20 °C)
Melting Point 121.6 °C
Molar Mass 144.17 g/mol
Bulk Density 300 kg/m3
Boiling Point 294.8 °C (1013 hPa)
Vapor Pressure  - 0.1 hPa (30 °C)
Flash Point 153 °C
Density 1.27 g/cm3 (20 °C)
Ignition Point 430 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR