Sodium, paraffin protect rods for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.22284.0100
8.22284.0250
8.22284.1000
8.22284.0100
8.22284.0250
8.22284.1000
EM8.22284.0100EA
126.4
CAD
EM8.22284.0100
EM8.22284.0250
EM8.22284.1000
CA8.22284.0100
CA8.22284.0250
CA8.22284.1000
Sodium, paraffin protect rods for synthesis, Sigma-Aldrich®
Sodium
Formula:
Na Boiling Pt: 881 °C (1013 hPa) Melting Pt: 97.8 °C Density: 0.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085307 CAS Number: 7440-23-5 EINECS: 231-132-9 UN: 1428 ADR: 4.3,I Merck Index: 13,08641 |
Specification Test Results
Solubility | (20 °C) explosion decomposition |
Melting Point | 97.8 °C |
Molar Mass | 22.99 g/mol |
Boiling Point | 889 °C (1013 hPa) |
Vapor Pressure | 1.6 hPa (400 °C) |
Density | 0.97 g/cm3 (20 °C) |
Ignition Point |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...