Benzyl alcohol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.22259.0100 8.22259.1000 8.22259.2500 8.22259.0100 8.22259.1000 8.22259.2500
EM8.22259.0100CS 458.2 CAD
EM8.22259.0100 EM8.22259.1000 EM8.22259.2500 CA8.22259.0100 CA8.22259.1000 CA8.22259.2500
Benzyl alcohol for synthesis, Sigma-Aldrich®
Benzyl alcohol
Formula: C₆H₅CH₂OH
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15.3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 40 g/l (20 °C)
Melting Point -15.3 °C
Molar Mass 108.14 g/mol
Boiling Point 205 °C (1013 hPa)
Vapor Pressure 0.03 hPa (20 °C)
Flash Point 101 °C DIN 51758
Refractive Index 1.54
Explosion Limit 1.3 - 13 %(V)
Density 1.05 g/cm3 (20 °C)
Ignition Point 435 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR