(-)-Quinine hydrochloride dihydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate

M-Clarity™ quality level = MQ200

8.22194.0025 8.22194.0100 8.22194.0025 8.22194.0100
EM8.22194.0025CS 17811.34 CAD
EM8.22194.0025 EM8.22194.0100 CA8.22194.0025 CA8.22194.0100
(-)-Quinine hydrochloride dihydrate for synthesis, Sigma-Aldrich®
(-)-Quinine hydrochloride dihydrate
Formula: C₂₀H₂₅ClN₂O₂·2H₂O
MW: 396.91 g/mol
Melting Pt: 158…160 °C
Flash Pt: 122
Storage Temperature: Ambient
MDL Number: MFCD00151248
CAS Number: 6119-47-7
EINECS: 205-001-1

Order Now

Specification Test Results

Solubility 62.5 g/l (20 °C)
Melting Point 115 °C (decomposition)
Molar Mass 396.91 g/mol
Bulk Density 310 kg/m3
Flash Point 122 °C
pH 6.0 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR