Tribenzylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.22163.0250 8.22163.9010 8.22163.0250 8.22163.9010
EM8.22163.0250CS 4883.78 CAD
EM8.22163.0250 EM8.22163.9010 CA8.22163.0250 CA8.22163.9010
Tribenzylamine for synthesis, Sigma-Aldrich®
Tribenzylamine
Formula: N(CH₂C₆H₅)₃
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 91…93 °C
Density: 0.9912 g/cm³ (95 °C)
Flash Pt: 65 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004773
CAS Number: 620-40-6
EINECS: 210-638-3
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c45b19a7c75302421b18150c43e96e03402f56d&vid=389de7f2863502b374023e1a04841d39e89717ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 91 - 93 °C
Molar Mass 287.4 g/mol
Boiling Point 380 - 390 °C
pH 6.5 - 9.0 (10 g/l, H2O, 20 °C) suspension


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR