6-Propyl-2-thiouracil for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.22137.0025 8.22137.0100 8.22137.0025 8.22137.0100
EM8.22137.0025CS 7256.94 CAD
EM8.22137.0025 EM8.22137.0100 CA8.22137.0025 CA8.22137.0100
6-Propyl-2-thiouracil for synthesis, Sigma-Aldrich®
6-Propyl-2-thiouracil
Formula: C₇H₁₀N₂OS
Boiling Pt: 448 °C (1013 hPa)
Melting Pt: 215…218 °C
MDL Number: MFCD00006041
CAS Number: 51-52-5
EINECS: 200-103-2
Merck Index: 12,08054

Order Now

Specification Test Results

Solubility 1.1 g/l (20 °C)
Melting Point 216 - 220 °C
Molar Mass 170.22 g/mol
Bulk Density 159 kg/m3
Flash Point 300 °C
pH 6 - 7.5 (5 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR