Phenanthrene zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


MQ200

8.22122.0100 8.22122.0250 8.22122.0100 8.22122.0250
EM8.22122.0100CS 4490.36 CAD
EM8.22122.0100 EM8.22122.0250 CA8.22122.0100 CA8.22122.0250
Phenanthrene zS, Sigma-Aldrich®
Phenanthrene
Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 340
Melting Pt: 97…99 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 153
Storage Temperature: Ambient
MDL Number: MFCD00001168
CAS Number: 85-01-8
EINECS: 201-581-5
UN: 3077
ADR: 9 III
Merck Index: 12,07354
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92527d856e1fe8582e54bf3812e1580ee0b91b05&vid=d8ec397dac001e49a7958f12aacb577cd11954da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 1.15 mg/l (25 °C)
Melting Point 97 - 100 °C
Molar Mass 178.23 g/mol
Bulk Density 690 kg/m3
Boiling Point 336 °C (1013 hPa)
Vapor Pressure 0.00012 hPa (20 °C)
Flash Point 171 °C
Ignition Point 450 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR