n-Nonadecane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: N-nonadecane

M-Clarity™ quality level = MQ200

8.22110.0025 8.22110.0100 8.22110.0025 8.22110.0100
EM8.22110.0025CS 5046.52 CAD
EM8.22110.0025 EM8.22110.0100 CA8.22110.0025 CA8.22110.0100
n-Nonadecane for synthesis, Sigma-Aldrich®
n-Nonadecane
Formula: H₃C(CH₂)₁₇CH₃
MW: 268.53 g/mol
Boiling Pt: 329.7 °C (1013 hPa)
Melting Pt: 32.5 °C
Density: 0.7845 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009012
CAS Number: 629-92-5
EINECS: 211-116-8

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 31 - 34 °C
Molar Mass 268.52 g/mol
Boiling Point 329 - 331 °C
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 100 °C
Refractive Index 1.4211 (70 °C, 589 nm)
Explosion Limit 0.6 - 6.5 %(V)
Density 0.79 g/cm3 (20 °C)
Ignition Point 205 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR