5-Methyl-2-hexanone ≥98% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.22093.0100 8.22093.1000 8.22093.9046 8.22093.0100 8.22093.1000 8.22093.9046
EM8.22093.0100CS 1488.36 CAD
EM8.22093.0100 EM8.22093.1000 EM8.22093.9046 CA8.22093.0100 CA8.22093.1000 CA8.22093.9046
5-Methyl-2-hexanone ≥98% (by GC) for synthesis, Sigma-Aldrich®
5-Methyl-2-hexanone
Formula: C₇H₁₄O
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: –74 °C
Density: 0.81 g/cm3 (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00008950
CAS Number: 110-12-3
EINECS: 203-737-8
UN: 2302
ADR: 3,III

Order Now

Specification Test Results

Solubility 5.4 g/l (20 °C)
Melting Point -74 °C
Molar Mass 114.19 g/mol
Boiling Point 144 °C (1013 hPa)
Vapor Pressure 5.3 hPa (20 °C)
Flash Point 43 °C
Refractive Index 1.4078 (20 °C, 589 nm)
Explosion Limit 1.4 %(V)
Density 0.81 g/cm3 (20 °C)
Ignition Point 455 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR