L(+)-Lithium lactate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Lithium (S)-2-hydroxypropanoate, (S)-(+)-Lactic acid lithium salt, (S)-2-Hydroxypropionic acid Lithium salt, Lithium (S)-2-Hydroxypropionate, L(+)-Lactic acid lithium salt, (S)-(+)-2-Hydroxypropanoic acid lithium salt, (S)-(+)-2-Hydroxypropionic acid lithium salt, Lithium L-Lactate, L-Lactic acid Lithium salt

M-Clarity™ quality level = MQ200

8.22084.0100 8.22084.0250 8.22084.0100 8.22084.0250
EM8.22084.0100CS 4134.86 CAD
EM8.22084.0100 EM8.22084.0250 CA8.22084.0100 CA8.22084.0250
L(+)-Lithium lactate for synthesis, Sigma-Aldrich®
L(+)-Lithium lactate
Formula: C₃H₅LiO₃
MW: 96.01 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00065512
CAS Number: 27848-80-2
EINECS: 248-692-5

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 96.01 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR