Benzyltrimethylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium chloride

M-Clarity™ quality level = MQ200

8.21961.0005 8.21961.0250 8.21961.0005 8.21961.0250
EM8.21961.0005CS 55966.76 CAD
EM8.21961.0005 EM8.21961.0250 CA8.21961.0005 CA8.21961.0250
Benzyltrimethylammonium chloride for synthesis, Sigma-Aldrich®
Benzyltrimethylammonium chloride
Formula: C₁₀H₁₆ClN
Melting Pt: 238 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011782
CAS Number: 56-93-9
EINECS: 200-300-3
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 800 g/l (20 °C)
Melting Point 243 °C
Molar Mass 185.7 g/mol
Bulk Density 135 °C
Boiling Point  - 0.0001 hPa (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR