Benzenesulfonamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21959.0100 8.21959.0500 8.21959.0100 8.21959.0500
EM8.21959.0100CS 1022.26 CAD
EM8.21959.0100 EM8.21959.0500 CA8.21959.0100 CA8.21959.0500
Benzenesulfonamide for synthesis, Sigma-Aldrich®
Benzenesulphonamide
Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Melting Pt: 149…152 °C
MDL Number: MFCD00007930
CAS Number: 98-10-2
EINECS: 202-637-1

Order Now

Specification Test Results

Solubility 4.3 g/l (16 °C)
Melting Point 149 - 152 °C
Molar Mass 157.18 g/mol
Bulk Density 700 kg/m3
Flash Point 250 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR