2-Ethylhexyl phosphate (mono and di ester mixture) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Phosphoric Acid Octyl Ester, Octyl Phosphate, Phosphoric Acid 2-Ethylhexyl Ester

M-Clarity™ quality level = MQ200

8.21943.0100 8.21943.1000 8.21943.0100 8.21943.1000
EM8.21943.0100CS 1027 CAD
EM8.21943.0100 EM8.21943.1000 CA8.21943.0100 CA8.21943.1000
2-Ethylhexyl phosphate (mono and di ester mixture) for synthesis, Sigma-Aldrich®
2-Ethylhexyl phosphate (mono and di ester mixture)
Storage Temperature: Ambient
MDL Number: MFCD00135884
CAS Number: 12645-31-7
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Boiling Point 180 °C (1013 hPa) decomposes
Flash Point 100 °C
Density 1.02 g/cm3 (20 °C)
pH 2.3 (10 g/l, H2O, 20 °C) (as an emulsion)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR