m-Toluic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.21902.0250
8.21902.1000
8.21902.0250
8.21902.1000
EM8.21902.0250CS
8713.7
CAD
EM8.21902.0250
EM8.21902.1000
CA8.21902.0250
CA8.21902.1000
m-Toluic acid for synthesis, Sigma-Aldrich®
m-Toluic acid
Formula:
C₈H₈O₂ Boiling Pt: 263 °C (1013 hPa) Melting Pt: 108…111 °C Density: 1.054 g/cm³ (20 °C) Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002523 CAS Number: 99-04-7 EINECS: 202-723-9 Merck Index: 12,09673 |
Specification Test Results
Solubility | 1 g/l (20 °C) |
Melting Point | 108 - 111 °C |
Molar Mass | 136.15 g/mol |
Bulk Density | 500 kg/m3 |
Boiling Point | 263 °C (1013 hPa) |
Flash Point | 150 °C |
pH | 3.4 (H2O, 20 °C) (saturated solution) |
Ignition Point | 500 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...