1,2-Epoxycyclohexane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Epoxycyclohexane, Cyclohexene oxide

M-Clarity™ quality level = MQ200

8.21862.0050 8.21862.0250 8.21862.0050 8.21862.0250
EM8.21862.0050CS 5444.68 CAD
EM8.21862.0050 EM8.21862.0250 CA8.21862.0050 CA8.21862.0250
1,2-Epoxycyclohexane for synthesis, Sigma-Aldrich®
1,2-Epoxycyclohexane
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 129 °C (1013 hPa)
Melting Pt: –27 °C
Density: 0.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005162
CAS Number: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: 8 (3),II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -27 °C
Molar Mass 98.14 g/mol
Boiling Point 131 - 132 °C (1013 hPa)
Vapor Pressure 12 hPa (20 °C)
Flash Point 24 °C
Refractive Index 1.4515 (20 °C, 589 nm)
Explosion Limit 1.2 - 6.5 %(V)
Density 0.97 g/cm3 (20 °C)
Ignition Point 335 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR