1-Octanethiol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21861.0100 8.21861.0500 8.21861.0100 8.21861.0500
EM8.21861.0100CS 1083.88 CAD
EM8.21861.0100 EM8.21861.0500 CA8.21861.0100 CA8.21861.0500
1-Octanethiol for synthesis, Sigma-Aldrich®
1-Octanethiol
Formula: C₈H₁₈S
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: –49 °C
Density: 0.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004912
CAS Number: 111-88-6
EINECS: 203-918-1
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.004 g/l (20 °C)
Melting Point -49 °C
Molar Mass 146.29 g/mol
Boiling Point 199 °C (1013 hPa)
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 70 °C
Refractive Index 1.4538 (20 °C, 589 nm)
Density 0.84 g/cm3 (20 °C)
Ignition Point 240 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR