9-Methylanthracene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21249.0001 8.21249.0005 8.21249.0001 8.21249.0005
EM8.21249.0001CS 92838.9 CAD
EM8.21249.0001 EM8.21249.0005 CA8.21249.0001 CA8.21249.0005
9-Methylanthracene for synthesis, Sigma-Aldrich®
9-Methylanthracene
Formula: C₁₅H₁₂
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 73…76 °C
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001261
CAS Number: 779-02-2
EINECS: 212-299-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.0003 mg/l (25 °C)
Melting Point 76 - 79 °C
Molar Mass 192.26 g/mol
Boiling Point 196 - 197 °C (16 hPa)
Refractive Index 1.6959 (99 °C, 589 nm)
Density 1.06 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR