Trifluoromethanesulfonic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Trifluoromethanesulphonic acid, Trifluoromethanesulfonic acid

M-Clarity™ quality level = MQ200

8.21166.0100 8.21166.0025 8.21166.0100 8.21166.1000 8.21166.0025 8.21166.1000
CA8.21166.0100EA 378 CAD
CA8.21166.0100 EM8.21166.0025 EM8.21166.0100 EM8.21166.1000 CA8.21166.0025 CA8.21166.1000
Trifluoromethanesulfonic acid for synthesis, Sigma-Aldrich®
Trifluoromethanesulphonic acid
Formula: CHF₃O₃S
MW: 150.08 g/mol
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1.69 g/cm³ (20 °C)
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble,(caution ! development of heat)
Molar Mass 150.08 g/mol
Boiling Point 162 °C (1013 hPa)
Vapor Pressure 10 hPa (55 °C)
Density 1.71 g/cm3 (20 °C)
pH  - 1 (H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR