4-Toluenethiol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.21130.0100
8.21130.0250
8.21130.0100
8.21130.0250
EM8.21130.0100CS
6244.16
CAD
EM8.21130.0100
EM8.21130.0250
CA8.21130.0100
CA8.21130.0250
4-Toluenethiol for synthesis, Sigma-Aldrich®
4-Toluenethiol
Formula:
C₇H₈S Boiling Pt: 195 °C (1013 hPa) Melting Pt: 39…42 °C Density: 1.022 g/cm³ (51 °C) Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004851 CAS Number: 106-45-6 EINECS: 203-399-1 UN: 2811 ADR: 6.1,II Merck Index: 12,09461 |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | 39 - 42 °C |
Molar Mass | 124.21 g/mol |
Boiling Point | 195 °C (1013 hPa) |
Flash Point | 70 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...