Titanium(IV) propanolate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Tetrapropyl orthotitanate, Titanium (IV) propoxide, Titanium tetrapropanolate

M-Clarity™ quality level = MQ200

8.21107.0100 8.21107.0500 8.21107.0100 8.21107.0500
EM8.21107.0100CS 1298.76 CAD
EM8.21107.0100 EM8.21107.0500 CA8.21107.0100 CA8.21107.0500
Titanium(IV) propanolate for synthesis, Sigma-Aldrich®
Titanium(IV) propanolate
Formula: C₁₂H₂₈O₄Ti
MW: 284.23 g/mol
Boiling Pt: 137
Density: 1.033 g/cm³ (20 °C)
MDL Number: MFCD00009365
CAS Number: 3087-37-4
EINECS: 221-411-3
UN: 2413
ADR: 3 III

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 284.22 g/mol
Boiling Point 170 °C (36 hPa)
Flash Point 38 °C
Refractive Index 1.4962 (25 °C, 589 nm)
Density 1.05 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR