N,N,N',N'-Tetramethyl-p-phenylenediamine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.21101.0005
8.21101.0025
8.21101.0005
8.21101.0025
EM8.21101.0005CS
111827.66
CAD
EM8.21101.0005
EM8.21101.0025
CA8.21101.0005
CA8.21101.0025
N,N,N',N'-Tetramethyl-p-phenylenediamine for synthesis, Sigma-Aldrich®
N,N,N',N'-Tetramethyl-p-phenylenediamine
Formula:
C₁₀H₁₆N₂ Boiling Pt: 260 °C (1013 hPa) Melting Pt: 48…50 °C Density: 1.08 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008309 CAS Number: 100-22-1 EINECS: 202-831-6 UN: 2811 ADR: 6.1,II Merck Index: 12,09367 |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | 48 - 50 °C |
Molar Mass | 164.24 g/mol |
Boiling Point | 260 °C |
Flash Point |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...