Ammonium 1-pyrrolidinecarbodithioate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.21100.0025 8.21100.0100 8.21100.0025 8.21100.0100
EM8.21100.0025CS 13719.14 CAD
EM8.21100.0025 EM8.21100.0100 CA8.21100.0025 CA8.21100.0100
Ammonium 1-pyrrolidinecarbodithioate for synthesis, Sigma-Aldrich®
Ammonium 1-pyrrolidinecarbodithioate
Formula: C₅H₁₂N₂S₂
MDL Number: MFCD00012720
CAS Number: 5108-96-3
EINECS: 225-834-4

Order Now

Specification Test Results

Solubility 190 g/l (20 °C)
Melting Point 146 - 151 °C
Molar Mass 164.28 g/mol
Bulk Density 400 kg/m3
pH 6 - 7 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR