Titanium(IV) butanolate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Titanium (IV) butoxide, Titanium tetrabutanolate, Tetra-n-butyl orthotitanate, Titanium (IV) n-butoxide

M-Clarity™ quality level = MQ200

8.21084.0100 8.21084.0500 8.21084.9010 8.21084.9040 8.21084.0100 8.21084.0500 8.21084.9010 8.21084.9040
EM8.21084.0100CS 1038.06 CAD
EM8.21084.0100 EM8.21084.0500 EM8.21084.9010 EM8.21084.9040 CA8.21084.0100 CA8.21084.0500 CA8.21084.9010 CA8.21084.9040
Titanium(IV) butanolate for synthesis, Sigma-Aldrich®
Titanium(IV) butanolate
Formula: C₁₆H₃₆O₄Ti
MW: 340.34 g/mol
Boiling Pt: 310
Melting Pt: -55
Density: 0.99 g/cm3 (20 °C)
Flash Pt: 47
Storage Temperature: Refrigerator
MDL Number: MFCD00009433
CAS Number: 5593-70-4
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Solubility (20 °C) (slow decomposition)
Melting Point -40 °C
Molar Mass 340.32 g/mol
Boiling Point 310 - 314 °C (1013 hPa)
Vapor Pressure 5.6 hPa (20 °C)
Flash Point 47 °C
Explosion Limit 2.0 - 12.0 %(V)
Density 0.99 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR