Titanium(IV) ethanolate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Titanium (IV) ethoxide, Titanium tetraethanolate, Tetraethyl orthotitanate

M-Clarity™ quality level = MQ200

8.21083.0250 8.21083.1000 8.21083.0250 8.21083.1000
EM8.21083.0250CS 2249.92 CAD
EM8.21083.0250 EM8.21083.1000 CA8.21083.0250 CA8.21083.1000
Titanium(IV) ethanolate for synthesis, Sigma-Aldrich®
Titanium(IV) ethanolate
Formula: C₈H₂₀O₄Ti
MW: 228.12 g/mol
MDL Number: MFCD00009071
CAS Number: 3087-36-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) Hydrolysis
Melting Point -40 °C
Molar Mass 228.15 g/mol
Boiling Point 150 - 155 °C (16 hPa)
Flash Point 33 °C
Refractive Index 1.5043 (20 °C, 589 nm)
Density 1.08 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR