(S)-(-)-α-Terpineol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21078.0005 8.21078.0250 8.21078.1000 8.21078.0005 8.21078.0250 8.21078.1000
EM8.21078.0005CS 3535.6 CAD
EM8.21078.0005 EM8.21078.0250 EM8.21078.1000 CA8.21078.0005 CA8.21078.0250 CA8.21078.1000
(S)-(-)-α-Terpineol for synthesis, Sigma-Aldrich®
(S)-(-)-α-Terpineol
Formula: C₁₀H₁₈O
MDL Number: MFCD00075926
CAS Number: 10482-56-1

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 32 - 34 °C
Molar Mass 154.25 g/mol
Boiling Point 215 - 217 °C (1013 hPa)
Flash Point 90 °C
Refractive Index 1.4813 (20 °C, 589 nm)
Density 0.935 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR