o-Terphenyl for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
2-Phenylbiphenyl, 1,2-Terphenyl, 2-Phenyl-1,1'-biphenyl, 1,2-Diphenylbenzene, 1,1:2,1-Terphenyl
M-Clarity™ quality level = MQ200
8.21077.0025
8.21077.0100
8.21077.0025
8.21077.0100
EM8.21077.0025CS
5311.96
CAD
EM8.21077.0025
EM8.21077.0100
CA8.21077.0025
CA8.21077.0100
o-Terphenyl for synthesis, Sigma-Aldrich®
o-Terphenyl
Formula:
C₁₈H₁₄ MW: 230.31 g/mol |
MDL Number:
MFCD00003055 CAS Number: 84-15-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Solubility |
|
Melting Point | 55 - 57 °C |
Molar Mass | 230.3 g/mol |
Boiling Point | 332 °C (1013 hPa) |
Flash Point | 163 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...