Phthaloyl dichloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Phthaloyl chloride, o-Phthaloyl dichloride
M-Clarity™ quality level = MQ200
8.21029.0050
8.21029.0250
8.21029.0050
8.21029.0250
EM8.21029.0050CS
4594.64
CAD
EM8.21029.0050
EM8.21029.0250
CA8.21029.0050
CA8.21029.0250
Phthaloyl dichloride for synthesis, Sigma-Aldrich®
Phthaloyl dichloride
Formula:
C₈H₄Cl₂O₂ MW: 203.02 g/mol |
MDL Number:
MFCD00000666 CAS Number: 88-95-9 UN: 3265 ADR: 8 II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 16 °C |
Molar Mass | 203.03 g/mol |
Boiling Point | 281 °C |
Flash Point |
|
Refractive Index | 1.5684 (20 °C, 589 nm) |
Density | 1.41 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...