Triethyl orthoacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1,1,1-Triethoxyethane

M-Clarity™ quality level = MQ200

8.20934.0100 8.20934.0500 8.20934.0100 8.20934.0500
EM8.20934.0100CS 864.26 CAD
EM8.20934.0100 EM8.20934.0500 CA8.20934.0100 CA8.20934.0500
Triethyl orthoacetate for synthesis, Sigma-Aldrich®
Triethyl orthoacetate
Formula: C₈H₁₈O₃
MW: 162.23 g/mol
Boiling Pt: 142
Flash Pt: 32
MDL Number: MFCD00009223
CAS Number: 78-39-7
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 162.22 g/mol
Boiling Point 142 °C (1013 hPa)
Flash Point 32 °C
Refractive Index 1.3958 (20 °C, 589 nm)
Explosion Limit 0.9 - 7.0 %(V)
Density 0.89 g/cm3 (20 °C)
Ignition Point 455 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR