3,5-Dihydroxytoluene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20933.0005 8.20933.0025 8.20933.0100 8.20933.0005 8.20933.0025 8.20933.0100
EM8.20933.0005CS 44519.66 CAD
EM8.20933.0005 EM8.20933.0025 EM8.20933.0100 CA8.20933.0005 CA8.20933.0025 CA8.20933.0100
3,5-Dihydroxytoluene for synthesis, Sigma-Aldrich®
3,5-Dihydroxytoluene
Formula: C₇H₈O₂
MDL Number: MFCD00002291
CAS Number: 504-15-4

Order Now

Specification Test Results

Solubility 80 g/l (20 °C)
Melting Point 109 °C
Molar Mass 124.14 g/mol
Boiling Point 147 °C (7 hPa)
Flash Point 159 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR