(±)-2-Octanol ≥98% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.20925.0100
8.20925.1000
8.20925.2500
8.20925.9026
8.20925.0100
8.20925.1000
8.20925.2500
8.20925.9026
EM8.20925.0100CS
883.22
CAD
EM8.20925.0100
EM8.20925.1000
EM8.20925.2500
EM8.20925.9026
CA8.20925.0100
CA8.20925.1000
CA8.20925.2500
CA8.20925.9026
(±)-2-Octanol ≥98% (by GC) for synthesis, Sigma-Aldrich®
(±)-2-Octanol
Formula:
C₈H₁₈O Boiling Pt: 177…182 °C (1013 hPa) Melting Pt: –38 °C Density: 0.82 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004591 CAS Number: 123-96-6 UN: 1993 ADR: 3,III |
Specification Test Results
Solubility | 1.12 g/l (25 °C) |
Melting Point | -38 °C |
Molar Mass | 130.23 g/mol |
Boiling Point | 177 - 182 °C (1013 hPa) |
Vapor Pressure | 0.25 hPa (20 °C) |
Flash Point | 71 °C |
Refractive Index | 1.4234 (20 °C, 589 nm) |
Explosion Limit | 0.80 - 7.40 %(V) |
Density | 0.82 g/cm3 (20 °C) |
Ignition Point | 255 - 260 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...