4'-Nitroacetanilide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
N-(4-Nitrophenyl)acetamide
M-Clarity™ quality level = MQ200
8.20880.0025
8.20880.0100
8.20880.0025
8.20880.0100
EM8.20880.0025CS
527.72
CAD
EM8.20880.0025
EM8.20880.0100
CA8.20880.0025
CA8.20880.0100
4'-Nitroacetanilide for synthesis, Sigma-Aldrich®
4'-Nitroacetanilide
Formula:
C₈H₈N₂O₃ MW: 180.16 g/mol |
MDL Number:
MFCD00007303 CAS Number: 104-04-1 |
Specification Test Results
Solubility | 2.2 g/l (25 °C) |
Melting Point | 212 - 215 °C |
Molar Mass | 180.16 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...