1-Methylnaphthalene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: α-Methylnaphthalene

M-Clarity™ quality level = MQ200

8.20809.0005 8.20809.0250 8.20809.1000 8.20809.0005 8.20809.0250 8.20809.1000
EM8.20809.0005CS 4648.36 CAD
EM8.20809.0005 EM8.20809.0250 EM8.20809.1000 CA8.20809.0005 CA8.20809.0250 CA8.20809.1000
1-Methylnaphthalene for synthesis, Sigma-Aldrich®
1-Methylnaphthalene
Formula: C₁₁H₁₀
MW: 142.2 g/mol
MDL Number: MFCD00004034
CAS Number: 90-12-0
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.026 g/l (25 °C)
Molar Mass 142.20 g/mol
Boiling Point 244 - 245 °C (1013 mbar)
Vapor Pressure 2 hPa (25 °C)
Flash Point 82 °C
Explosion Limit 0.7 - 6.5 %(V)
Density 1.02 g/cm3 (20 °C)
Ignition Point 529 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR