Ethyl methanesulfonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20774.0005 8.20774.0025 8.20774.0005 8.20774.0025
EM8.20774.0005CS 5714.86 CAD
EM8.20774.0005 EM8.20774.0025 CA8.20774.0005 CA8.20774.0025
Ethyl methanesulfonate for synthesis, Sigma-Aldrich®
Ethyl methanesulphonate
Formula: C₃H₈O₃S
MDL Number: MFCD00007559
CAS Number: 62-50-0
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 50 - 100 g/l (27 °C)
Molar Mass 124.16 g/mol
Boiling Point 213 - 214 °C (1013 hPa)
Vapor Pressure 0.27 hPa (25 °C)
Flash Point 100 °C
Refractive Index 1.418 (20 °C, 589 nm)
Density 1.206 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR