(±)-Mercaptosuccinic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.20763.0100 8.20763.5000 8.20763.0100 8.20763.5000
EM8.20763.0100CS 1467.82 CAD
EM8.20763.0100 EM8.20763.5000 CA8.20763.0100 CA8.20763.5000
(±)-Mercaptosuccinic acid for synthesis, Sigma-Aldrich®
(±)-Mercaptosuccinic acid
Formula: C₄H₆O₄S
MDL Number: MFCD00004860
CAS Number: 70-49-5

Order Now

Specification Test Results

Solubility 150 g/l (20 °C)
Melting Point 150 - 153 °C
Molar Mass 150.15 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR