Iodobenzene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Phenyliodide
M-Clarity™ quality level = MQ200
8.20730.0100
8.20730.0250
8.20730.0100
8.20730.0250
EM8.20730.0100CS
2565.92
CAD
EM8.20730.0100
EM8.20730.0250
CA8.20730.0100
CA8.20730.0250
Iodobenzene for synthesis, Sigma-Aldrich®
Iodobenzene
Formula:
C₆H₅I MW: 204.01 g/mol |
MDL Number:
MFCD00001029 CAS Number: 591-50-4 |
Specification Test Results
Solubility | 0.34 g/l (30 °C) (experimental) |
Melting Point | -30 °C |
Molar Mass | 204.01 g/mol |
Boiling Point | 187 - 189 °C |
Vapor Pressure | 1.41 hPa (25 °C) |
Flash Point | 77 °C |
Refractive Index | 1.6198 (20 °C, 589 nm) |
Density | 1.83 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...