Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-Hydroxyethanesulfonic acid sodium salt, Sodium isethionate, Sodium hydroxyethyl sulphonate, Sodium hydroxyethyl sulfonate, 2-Hydroxyethanesulphonic acid sodium salt

M-Clarity™ quality level = MQ200

8.20708.0100 8.20708.1000 8.20708.9050 8.20708.0100 8.20708.1000 8.20708.9050
EM8.20708.0100CS 10334.78 CAD
EM8.20708.0100 EM8.20708.1000 EM8.20708.9050 CA8.20708.0100 CA8.20708.1000 CA8.20708.9050
Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) for synthesis, Sigma-Aldrich®
Isethionic acid sodium salt (Sodium 2-hydroxyethanesulphonate)
Formula: C₂H₅NaO₄S
MW: 148.11 g/mol
Melting Pt: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00007534
CAS Number: 1562-00-1

Order Now

Specification Test Results

Solubility 650 g/l (20 °C)
Melting Point 192 - 194 °C
Molar Mass 148.11 g/mol
Bulk Density 800 kg/m3
pH 7.0 - 11.0 (20 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR